4-Bromobenzenediazonium tetrafluoroborate, 96%
4-Bromobenzenediazonium tetrafluoroborate, 96%
4-Bromobenzenediazonium tetrafluoroborate, 96%
Thermo Scientific Chemicals

4-Bromobenzenediazonium tetrafluoroborate, 96%

CAS: 673-40-5 | C6H4BBrF4N2 | 270.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number B25670.14
also known as B25670-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS673-40-5
IUPAC Name4-bromobenzene-1-diazonium; tetrafluoroboranuide
Molecular FormulaC6H4BBrF4N2
InChI KeyFXTCQUCYDJZGGU-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown or pale pink
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
RUO – Research Use Only

General References:

  1. Bo Cui; Jing-Ying Gu; Ting Chen; Hui-Juan Yan; Dong Wang; Li-Jun Wan. Solution effect on diazonium-modified Au(111): reactions and structures. Langmuir. 2013, 29 (9), 2955-2960.
  2. Sylvain Darses; Mathieu Pucheault and Jean-Pierre Genêt. Efficient access to perfluoroalkylated aryl compounds by Heck reaction. European Journal of Organic Chemistry. 2001, 2001 (6), 1121-1128.