3-Bromobenzenesulfonamide, 97%
3-Bromobenzenesulfonamide, 97%
3-Bromobenzenesulfonamide, 97%
3-Bromobenzenesulfonamide, 97%
Thermo Scientific Chemicals

3-Bromobenzenesulfonamide, 97%

CAS: 89599-01-9 | C6H6BrNO2S | 236.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25681.03
also known as B25681-03
Price (EUR)
32,20
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
32,20
Each
Add to cart
Chemical Identifiers
CAS89599-01-9
IUPAC Name3-bromobenzene-1-sulfonamide
Molecular FormulaC6H6BrNO2S
InChI KeyMUBJNMWVQGHHLG-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
Assay (GC)≥96.0%
Melting Point (clear melt)151.0-161.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only