7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%
7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%
7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%
7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%
Thermo Scientific Chemicals

7-Bromo-1,2,3,4-tetrahydroisoquinoline, 97%

CAS: 17680-55-6 | C9H10BrN | 212.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25712.03
also known as B25712-03
Price (EUR)
73,10
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
73,10
Each
Add to cart
Chemical Identifiers
CAS17680-55-6
IUPAC Name7-bromo-1,2,3,4-tetrahydroisoquinoline
Molecular FormulaC9H10BrN
InChI KeyOYODEQFZAJVROF-UHFFFAOYSA-N
SMILESBrC1=CC2=C(CCNC2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
7-Bromo-1,2,3,4-tetrahydroisoquinoline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1,2,3,4-tetrahydroisoquinoline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. George J. Quallich; Teresa W. Makowski; Andrew F. Sanders; Frank J. Urban and Enrique Vazquez. Synthesis of 1,2,3,4-Tetrahydroisoquinolines Containing Electron-Withdrawing Groups. J. Org. Chem.. 1998, 63 (12), 4116-4119.
  2. Richard A Glennon; Richard Young; Jagadeesh B Rangisetty. Further characterization of the stimulus properties of 5,6,7,8-tetrahydro-1,3-dioxolo[4,5-g]isoquinoline. Pharmacology Biochemistry and Behavior. 2002, 72 (1-2), 379-387.