Is the PowerEase Touch Power Supply (Cat. No. PS0351) compatible with the Owl gel Systems?
Yes, it is compatible with the Owl Gel Systems. The following ones can be used with the PowerEase power supplies with no need for adaptors:
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
- Owl EasyCast B1 Mini Gel Electrophor. Sys. (Cat. No. B1-BP)
- Owl A2 Large Gel Sys. (Cat. No. A2)
- Owl EasyCast B3 Gel Sys. with Built-In Recirculat. (Cat. No. B3).
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.