Biotin-XX Microscale Protein Labeling Kit - FAQs
Biotin-XX Microscale Protein Labeling Kit - FAQs
View additional product information for Biotin-XX Microscale Protein Labeling Kit - FAQs (B30010)
1 product FAQs found
Can I use 50 μg of protein with Fluorescent Protein Labeling Kits?
No. We recommend using 1 mg of protein with Fluorescent Protein Labeling Kits. For smaller protein sample sizes, we recommend using Microscale Protein Labeling kits which are optimized for 20-100 µg of protein.