What dyes are included in the BrainStain Imaging Kit?
The BrainStain Imaging Kit enables three-color staining of myelin, neurons, and nuclei in brain cryosections in a single 20-minute staining step. It includes FluoroMyelin Green dye, NeuroTrace 530/615 red fluorescent Nissl stain, and DAPI for staining nuclei.