BacLight™ Green Bacterial Stain - FAQs
BacLight™ Green Bacterial Stain - FAQs
View additional product information for BacLight™ Green Bacterial Stain - FAQs (B35000)
2 product FAQs found
What bacterial parameters can I look at by flow cytometry?
You can stain bacteria with a general stain such as BacLight Green Bacterial Stain (Cat. No. B35000) or BacLight Red Bacterial Stain (Cat. No. B35001). You can look at gram character (Cat. No. L7005), cell viability (Cat. Nos. L7007, L7012, and L13152), cell count (Cat. Nos. L34856 and B7277), and cell vitality. Cell vitality can be measured by membrane potential (Cat. No. B34950) or by metabolism (Cat. Nos. B34954 and B34956).
What optical filter do your recommend with the BacLight Green Bacterial Stain?
We recommend filters suitable for fluorescein/FITC with the BacLight Green kit.