Thermo Scientific™
Shandon™ PolarHeat™ Sheets
 Related applications:
Anatomical Pathology
Heat paraffin in the microwave with break-through Thermo Scientific™ Shandon™ PolarHeat™ sheets.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Eliminates pre-heating
  • Disseminates microwaves evenly through paraffin
Documents
Manuals & protocols