Steriking™ Gusseted Pouch - FAQs
Steriking™ Gusseted Pouch - FAQs
View additional product information for Steriking™ Gusseted Pouch - FAQs (B36)
1 product FAQs found
What are white precipitates in the Thermo Scientific 10X T4 DNA Ligase Buffer that appear upon thawing?
The white precipitates are DTT in the buffer which may appear after freezing. To redissolve DTT, warm up the buffer to 37 degrees C by holding the vial between your palms, or incubating in a water bath for a few minutes, and mix well. The formation of DTT precipitates does not influence the quality of the buffer. The 10X T4 DNA Ligase Buffer is also available separately (Cat. No. B69).