EquiPhi29™ DNA Polymerase Reaction Buffer (10X)
Thermo Scientific™

EquiPhi29™ DNA Polymerase Reaction Buffer (10X)

This reaction buffer is for use with EquiPhi29 DNA Polymerase. Buffer is included with purchase of the polymerase, but additional buffer is made available here to help accommodate experimental needs.
Have Questions?
카탈로그 번호수량
B395 x 1 mL
카탈로그 번호 B39
제품 가격(KRW)
46,000
온라인 행사
Ends: 31-Dec-2026
52,000
할인액 6,000 (12%)
Each
수량:
5 x 1 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
46,000
온라인 행사
Ends: 31-Dec-2026
52,000
할인액 6,000 (12%)
Each

This reaction buffer is for use with EquiPhi29 DNA Polymerase. Buffer is included with purchase of the polymerase, but additional buffer is made available here to help accommodate experimental needs.

For Research Use Only. Not for use in diagnostic procedures.

사양
농도10X
배송 조건Dry Ice
용도(애플리케이션)Isothermal Amplification
수량5 x 1 mL
Unit SizeEach
구성 및 보관

• 10X Reaction buffer for EquiPhi29 DNA Polymerase, 5 x 1.0 mL

Store at -5°C to -30°C.