BODIPY™ 665/676 (Lipid Peroxidation Sensor)
Invitrogen™
BODIPY™ 665/676 (Lipid Peroxidation Sensor)
BODIPY 665/676 is a lipophilic probe used to detect lipid peroxidation. It exhibits a change in fluorescence after interaction with peroxyl radicals within cells.
Have Questions?
카탈로그 번호수량
B39325 mg
카탈로그 번호 B3932
제품 가격(KRW)
360,000
온라인 행사
Ends: 30-Sep-2026
411,000
할인액 51,000 (12%)
Each
수량:
5 mg
제품 가격(KRW)
360,000
온라인 행사
Ends: 30-Sep-2026
411,000
할인액 51,000 (12%)
Each
BODIPY 665/676 is a lipophilic probe used to detect lipid peroxidation. It exhibits a change in fluorescence after interaction with peroxyl radicals within cells. We recommend dissolving BODIPY 665/676 in high-quality anhydrous DMSO to attain a stock concentration of 1 to 10 mM.
Lipid peroxidation is the oxidative degradation of cellular lipids by free radicals from reactive oxygen species, which can degrade lipids containing carbon-carbon double bonds such as phospholipids and polyunsaturated fatty acids. Oxidation of lipids leads to the generation of lipid peroxides and can cause damage to cell membranes, resulting in changes to signal transduction pathways and eventually leading to cell death. Lipid peroxidation is involved in apoptosis and one of the main contributors to ferroptosis, an iron-dependent, non-apoptotic form of cell death. Oxidative stress from lipid peroxidation plays a role in aging, as well as pathologies such as cancer, atherosclerosis, and neurodegenerative diseases.
For Research Use Only. Not for use in diagnostic procedures.
사양
색상Far-red
용도(애플리케이션)Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
수량5 mg
배송 조건Room Temperature
타겟 기능Lipid peroxidation
검출 방법Fluorescence
Excitation/Emission665/676 nm
IndicatorROS
제품라인BODIPY
제품 유형Lipid Sensor
Unit SizeEach
인용 및 참조 문헌 (8)
인용 및 참조 문헌
Abstract
Synthesis, chiroptical properties, and solid-state structure determination of two new chiral dipyrrin difluoroboryl chelates.
Authors:Gossauer A, Nydegger F, Kiss T, Sleziak R, Stoeckli-Evans H
Journal:J Am Chem Soc
PubMed ID:14871109
Two new types of optically active BODIPY fluorophores bearing chiral phenyl substituents either at the meso-position or at both external alpha-positions have been synthesized. Their chiroptical properties are strongly dependent both on the position of the chiral group and on the protonation of the chromophore. The solid-state structures of one ... More
Aggregation of puroindoline in phospholipid monolayers spread at the air-liquid interface.
Authors:Dubreil L, Vié V, Beaufils S, Marion D, Renault A
Journal:Biophys J
PubMed ID:14507728
Puroindolines, cationic and cystine-rich low molecular weight lipid binding proteins from wheat seeds, display unique foaming properties and antimicrobial activity. To unravel the mechanism involved in these properties, the interaction of puroindoline-a (PIN-a) with dipalmitoylphosphatidylcholine (DPPC) and dipalmitoylphosphatidylglycerol (DPPG) monolayers was studied by coupling Langmuir-Blodgett and imaging techniques. Compression isotherms ... More
Antioxidant activities of astaxanthin and related carotenoids.
Authors:Naguib YM
Journal:J Agric Food Chem
PubMed ID:10775364
The antioxidant activities of astaxanthin and related carotenoids have been measured by employing a newly developed fluorometric assay. This assay is based on 4,4-difluoro-3,5-bis(4-phenyl-1, 3-butadienyl)-4-bora-3a,4a-diaza-s-indacene (BODIPY 665/676) as an indicator; 2,2'-azobis-2,4-dimethylvaleronitrile (AMVN) as a peroxyl radical generator; and 6-hydroxy-2,5,7, 8-tetramethylchroman-2-carboxylic acid (Trolox) as a calibrator in an organic and liposomal ... More
Multiple C2 domain-containing transmembrane proteins promote lipid droplet biogenesis and growth at specialized endoplasmic reticulum subdomains.
Authors:
Journal:Mol Biol Cell
PubMed ID:33826368
Surface-associated lipid droplets: an intermediate site for lipid transport in human adipocytes?
Authors:
Journal:Adipocyte
PubMed ID:33108251