DRI Tramadol Drugs of Abuse Assay
Thermo Scientific™

DRI Tramadol Drugs of Abuse Assay

Developed for hospital laboratories, addiction treatment centers, and toxicology laboratories, the Thermo Scientific™ DRI™ Tramadol Drugs of Abuse Assay is an effective tool for the screening of tramadol abuse and misuse.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
B400003501 x 500 mL
B400003511 x 115mL
Catalog number B40000350
Price (CLP)
-
Quantity:
1 x 500 mL

Developed for hospital laboratories, addiction treatment centers, and toxicology laboratories, the Thermo Scientific™ DRI™ Tramadol Drugs of Abuse Assay is an effective tool for the screening tramadol abuse and misuse. Its ability to detect tramadol and its primary metabolites, O-Desmethyltramadol and N-Desmethyltramadol gives this assay a broader window of detection compared to competing products. This automated urine drug screening assay exhibits negligible cross-reactivity to other structurally related compounds. It is the only on-market 510(k) cleared assay that demonstrates a cross-reactivity with a low 100 ng/mL cutoff concentration.

  • Specifically detect tramadol and its primary metabolites O-Desmethyltramadol and N-Desmethyltramadol at physiologically relevant concentrations in human urine
  • Low cross-reactivity to non-tramadol compounds
  • High sensitivity with a cutoff concentration of 100 ng/mL
  • Superior assay precision
  • Excellent correlation to LC-MS/MS
  • Liquid, Ready-to Use
For In Vitro Diagnostic Use.  
Specifications
Control SetsDRI Tramadol Drugs of Abuse Control Kit
DescriptionDRI Tramadol Drugs of Abuse Assay
For Use With (Application)Enzyme Immunoassay Detection
FormatKit
Quantity1 x 500 mL
Sample TypeUrine
TypeUrine
Detectable AnalytesTramadol (TRA)
DoA CalibratorsNo
Unit SizeEach