SuperBoost™ tyramide signal amplification is the most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC),Read more
Have Questions?
Catalog Number
Quantity
B40931
150 Slides
Catalog number B40931
Price (USD)
789.65
Online exclusive
858.00
Save 68.35
Each
Quantity:
150 Slides
Price (USD)
789.65
Online exclusive
858.00
Save 68.35
Each
SuperBoost™ tyramide signal amplification is the most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH). SuperBoost kits combine the brightness of AlexaFluor™ dyes with the superior signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce a sensitivity 10-200 times greater than standard methods. For standout research, SuperBoost kits sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal.
SuperBoost kits are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a SuperBoost kit can be imaged using any type of microscope, producing high-resolution multiplex images. This particular kit features biotin XX tyramide with HRP-conjugated streptavidin. For detection and additional signal amplification, conjugated streptavidin is used to detect specifically deposited biotin XX .
Features of the SuperBoost kits include: • Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging • Simple protocol and detection using standard filters • Suitable for high-resolution multiplex images—co-label with DAPI, secondary antibodies, and other SuperBoost kits • Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
SuperBoost kits are based on the tyramide signal amplification system, which uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labeling of a target protein or nucleic acid sequence in situ. A typical ICC/IHC/ISH experiment using a SuperBoost kit requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments. SuperBoost kits offer superior specific signal intensity over background, so the protocol is easily optimized to detect specific signal in samples where high endogenous autofluorescence is observed.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeTyramide Kit
Quantity150 Slides
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
ConjugateBiotin-XX
Product LineSuperBoost
Unit SizeEach
Contents & Storage
1 kit sufficient for 150 microscope slides (18 x 18 mm), containing: Blocking buffer (1 X), 22.5 mL
HRP-conjugated streptavidin (1 X), 22.5 mL
Biotin-XX tyramide reagent
Hydrogen peroxide (stabilized 3% solution
Frequently asked questions (FAQs)
With a SuperBoost tyramide kit, I got excessive and non-specific labeling. What can I do to limit background and acquire a more localized labeling?
To limit background, we recommend performing a pre-blocking step with 3% H2O2 for 60 mins to inactivate endogenous peroxidases. To limit the localization of labeling, we recommend optimizing the final concentration of the primary and secondary antibodies and the dye-tyramide. You may also limit the incubation time of the dye-tyramide on the sample.
How are SuperBoost tyramide kits different from the original TSA labeling kits?
The SuperBoost tyramide kits utilize poly-HRP labeled antibodies. This provides a greater number of horseradish peroxidase (HRP) molecules per antibody. The original kits used antibodies and streptavidin that were directly conjugated with HRP and thus, limited the number per antibody or streptavidin.
Soluble amyloid-beta buffering by plaques in Alzheimer disease dementia versus high-pathology controls.
Authors:Esparza TJ, Gangolli M, Cairns NJ, Brody DL,
Journal:PLoS One
PubMed ID:29979775
An unanswered question regarding Alzheimer disease dementia (ADD) is whether amyloid-beta (Aß) plaques sequester toxic soluble Aß species early during pathological progression. We previously reported that the concentration of soluble Aß aggregates from patients with mild dementia was higher than soluble Aß aggregates from patients with modest Aß plaque burden ... More