SuperBoost™ Goat anti-Mouse Poly HRP is a component of SuperBoost™ tyramide signal amplification kits made available separately here. SuperBoost™ Goat자세히 알아보기
Have Questions?
카탈로그 번호
수량
B40961
150 slides
카탈로그 번호 B40961
제품 가격(KRW)
403,000
온라인 행사
Ends: 31-Dec-2026
473,000
할인액 70,000 (15%)
Each
수량:
150 slides
제품 가격(KRW)
403,000
온라인 행사
Ends: 31-Dec-2026
473,000
할인액 70,000 (15%)
Each
SuperBoost™ Goat anti-Mouse Poly HRP is a component of SuperBoost™ tyramide signal amplification kits made available separately here. SuperBoost™ Goat anti-Mouse Poly HRP can be used with any HRP sustrate such as tyramide, DAB for signal amplification and detection. The molar enzyme/antibody protein ratio has an average value of '4'. This reagent is provided at a ready-to-use 1X concentration, enough for 150 slides.
When used as directed, this reagent along with other SuperBoost reagents can be used for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH) with up to 200 times more sensitivity than standard methods. For standout research, SuperBoost reagents sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal. SuperBoost reagents are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a SuperBoost reagent can be imaged using any type of microscope, producing high-resolution multiplex images.
Features of the SuperBoost reagents include: • Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging • Simple protocol and detection using standard filters • Suitable for high-resolution multiplex images—co-label with TO-PRO™-3, secondary antibodies, and other SuperBoost kits • Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
For Research Use Only. Not for use in diagnostic procedures.
사양
제품라인SuperBoost
배송 조건Approved for shipment at Room Temperature or on Wet Ice
콘주게이트HRP
수량150 slides
Unit SizeEach
구성 및 보관
22.5 mL at 1X concentration in dropper bottle Store at 2-8°C. When stored as directed, the product is stable for 6 months after receiving.