Gel Runner Tank
Invitrogen™
Gel Runner Tank
The Gel Runner Tank is a replacement part for the Mini Gel Tank. A tank lid, tank base, and cassetteRead more
Have Questions?
Catalog NumberQuantity
B44786411 unit
Catalog number B4478641
Price (MXN)
4,507.51
Each
Quantity:
1 unit
Price (MXN)
4,507.51
Each
The Gel Runner Tank is a replacement part for the Mini Gel Tank. A tank lid, tank base, and cassette clamps are not included.
About the Mini Gel Tank
The Mini Gel Tank is compatible with a variety of Novex® gels, NuPAGE® gels, and Bolt® Bis-Tris Plus gels. Each Mini Gel Tank can accommodate up to two gels per run. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt® Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 unit
For Use With (Equipment)Bolt™ System
Gel CompatibilityBolt™ Bis-Tris Plus Gels
Gel SizeMini
Product LineBolt
TypeGel Runner Tank
Unit SizeEach
Contents & Storage
Runner Tank
Frequently asked questions (FAQs)
Can I purchase replacement parts for the Mini Blot Module?
Here are the replacement parts we offer for the Mini Blot Module:
- Mini Blot Module Gasket, Cat. No. B1001
- Gel Knife, Cat. No. EI9010
- Bolt Tank Base, Cat. No. B4478640
- Gel Runner Tank, Cat. No. B4478641
- Mini Gel Tank Lid, Cat. No. A25944
- Cassette Clamp, Right, Cat. No. B4478592
- Cassette Clamp, Left, Cat. No. B4478593
- Power Supply Adapters, Cat. No. ZA10001
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I purchase replacement parts for the Mini Gel Tank?
Here are the replacement parts we offer for the Mini Gel Tank:
Replacement part - Cat. No.
Gel Runner Tank - B4478641
Mini Gel Tank Lid - A25944
Mini Gel Tank Base - A25950
Cassette Clamp, left - A25946
Cassette Clamp, right - A25945
Cassette Clamp Cam Handle Set - A26732
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the components of the Mini Gel Tank?
The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Invitrogen Power Supply Adapters, and Gel Knife.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.