Thermo Scientific 50X TAE Buffer (Tris-acetate-EDTA) is used for electrophoresis of nucleic acids in agarose and polyacrylamide gels. You canRead more
Have Questions?
Catalog Number
Quantity
B49
1 L
Catalog number B49
Price (USD)
113.65
Online exclusive
134.00
Save 20.35
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
113.65
Online exclusive
134.00
Save 20.35
Each
Thermo Scientific 50X TAE Buffer (Tris-acetate-EDTA) is used for electrophoresis of nucleic acids in agarose and polyacrylamide gels. You can use this buffer for both genomic and large supercoiled DNA, and you can also use this as both a running and a gel preparation buffer.
Applications
• Electrophoresis of nucleic acids in agarose and polyacrylamide gels • Used both as a running buffer and as a gel preparation buffer • Filtered through a 0.22 μm membrane • Recommended for resolution of RNA and DNA fragments larger than 1500 bp, for genomic DNA and for large supercoiled DNA
Note TAE buffer has a relatively low buffering capacity. Therefore, periodic replacement of the buffer during prolonged electrophoresis is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeTAE Buffer
Quantity1 L
Concentration50X
Unit SizeEach
Frequently asked questions (FAQs)
What is the 1X buffer composition of your TAE electrophoresis buffer (Cat. No. B49)?
The 1X buffer composition is as follows: 40 mM Tris, 20 mM acetic acid, 1mM EDTA.