OptiMSe™ Software
Thermo Scientific™
OptiMSe™ Software
For fast sample screening, Thermo Scientific™ OptiMSe™ software empowers rapid data acquisition and analysis, provides confidence in your sample prep and optimizes Electron Microscopy utilization.
Have Questions?
Catalog NumberType
B51002092Software
Catalog number B51002092
Price (USD)
-
Type:
Software
OptiMSe software facilitates data analysis for fast screening of intact proteins and protein complexes. Its on-the-fly report enables sample selection for downstream cryo-em analysis. OptiMSe software is developed for use on native MS data collected via online buffer exchange.
OptiMSe software
  • Applies novel in-house developed algorithms for isotopically unresolved and smaller isotopically resolved protein species.
  • Algorithm and parameters are automatically selected based on user-defined protein target mass.
  • Processes data concurrently on existing instrument hardware, post run so that results are ready shortly after the last run is completed.
Specifications
DescriptionOptiMSe 1.0 ​software
For Use With (Equipment)Mass spectrometers
TypeSoftware
Unit SizeEach