Thermo Scientific™ Cindion™ Solid and Liquid Boats boats hold samples for use in combustion ion chromatography analysis using the Thermo Scientific™ Cindion™ Combustion IC System.
Have Questions?
Change view
Catalog Number
Type
B51006445
Liquids Boat
B51006443
Solids Boat
2 Options
Catalog number B51006445
Price (EUR)
-
Type:
Liquids Boat
Use these quartz sample boats to hold samples for insertion into the combustion tube for combustion-ion chromatography (C-IC) analysis. These boats are available for solid or liquid samples.
These boats are compatible with solid and liquid sample configurations of the Cindion Combustion IC System.
Boat is introduced into the combustion oven at a programmed rate
Specifications
Dimensions (D x W x H)2 cm x 16 cm x 2 cm
For Use With (Application)Combustion IC for Liquid Samples
For Use With (Equipment)Cindion Combustion IC System