Product Image
Thermo Scientific™

DreamTaq™ Buffers (10X)

Thermo Scientific 10X DreamTaq Buffer is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoColor
B71Green
B65Colorless
Número de catálogo B71
Precio (EUR)
27,87
Each
Color:
Green
Pedido a granel o personalizado
Precio (EUR)
27,87
Each
Thermo Scientific DreamTaq Buffer (10X), available in colorless and green formats, is specifically optimized for PCR using DreamTaq DNA Polymerase and DreamTaq Hot Start DNA Polymerase. It contains both KCl and (NH4)2SO4 to provide high specificity of primer annealing at a wide range of MgCl2 concentrations. 10X DreamTaq buffer includes 20 mM MgCl2

In addition, 10X DreamTaq Green Buffer includes a density reagent and two tracking dyes. The density reagent allows direct loading of PCR products on a gel. The blue dye (migrates with 3–5 kb DNA fragments in 1% agarose gel) and the yellow dye (migrates faster than 10 bp DNA fragments in 1% agarose gel) are included for monitoring electrophoresis progress. The dyes have excitation peaks at 424 nm and 615 nm.

Notes

  • For applications that require PCR product analysis by absorbance or fluorescence excitation, we recommend using the colorless DreamTaq Buffer.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Nombre del producto químico o materialTampón verde DreamTaq
ColorGreen
FormulaciónNonidet P40 etilfenolpoli(etilenogiycoleter) 9016-45-9 (<0,9 ), polietilenglicol 25322-68-3 (7-13 ), Sucrose 57-50-1 (10-30)
IncluyeContiene:
• Tampón verde DreamTaq de 10 x 1,25 ml (incluye 20 mM MgCl2)

Almacenar a -20 °C.
Almacenamiento recomendadoContiene:
• Tampón verde DreamTaq de 10 x 1,25 ml (incluye 20 mM MgCl2)

Almacenar a -20 °C.
Concentración10X
Para utilizar con (aplicación)Hot-start PCR
Cantidad4 x 1.25 mL
Unit SizeEach