Components for Vial Convenience Kits
Components for Vial Convenience Kits
Thermo Scientific™

Components for Vial Convenience Kits

Vials, caps and septa for use with Thermo Scientific™ National Vial Convenience Kits.
Have Questions?
Catalog NumberVolume (Metric)Color
B7921-VO20 mLAmber
Catalog number B7921-VO
Price (USD)
73.52
Pack of 100
Add to cart
Volume (Metric):
20 mL
Color:
Amber
Price (USD)
73.52
Pack of 100
Add to cart

Vials, caps and septa for use with Thermo Scientific™ National vial convenience kits.

Specifications
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
DescriptionAmber 20mL vials, 28 x 57mm
For Use With (Equipment)Thermo Scientific™ National Vial Convenience Kits
Height (Metric)57 mm
Product TypeVial
Volume (Metric) Total20 mL
Closure TypeScrew Cap
ColorAmber
Diameter (Metric) Outer28 mm
Product LineNational
Quantity100 Pk.
Volume (Metric)20 mL
Unit SizePack of 100