Product Image
Gibco™

High Five™ Cells in Express Five™ Medium

High Five™ Cells (BTI-TN-5B1-4) are a clonal isolate derived from the parental Trichoplusia ni cell line (cabbage looper ovary) andRead more
Have Questions?
Catalog NumberQuantity
B855023 x 106 cells
Catalog number B85502
Price (USD)
1,498.00
Each
Quantity:
3 x 106 cells
Price (USD)
1,498.00
Each
High Five™ Cells (BTI-TN-5B1-4) are a clonal isolate derived from the parental Trichoplusia ni cell line (cabbage looper ovary) and are commonly used for the expression of recombinant proteins using the Baculovirus Expression Vector System (BEVS). High Five™ Insect Cells are adapted to serum-free culture in Express Five™ SFM for maximum cell growth and recombinant gene expression. High Five™ Insect Cells (frozen in Express Five™ SFM) features:

• Recombinant protein expression from a variety of expression systems
• Up to ten-fold increase in secreted protein expression compared to Sf9 cells
• Good growth in adherent or suspension culture
• Quality and performance testing

Recombinant protein expression from a variety of expression systems
High levels of protein expression in High Five™ cells can be obtained using either the BaculoDirect™ Baculovirus Expression System, the Bac-to-Bac™ Baculovirus Expression System, or the InsectDirect™ Expression System.

Up to ten-fold increase in secreted protein expression compared to Sf9 cells
High Five™ cells are the best choice for expression of secreted protein in insect cells. They consistently express 5–10 times more protein that Sf9 cells.

Good growth in adherent or suspension culture
High Five ™ cells thaw into adherent culture, forming an irregular monolayer with plaques that may be difficult to isolate. Protocols for adherent growth can be found in Express Five™ SFM are available in the product manual. High Five™ cells can be adapted to suspension culture.

Quality and performance testing
Each lot of High Five ™ cells is tested for cell growth and viability post-recovery from cryopreservation.

Caution: Handle as potentially biohazardous material under at least Biosafety Level 2 containment. This product contains Dimethyl Sulfoxide (DMSO), a hazardous material. Review the Material Safety Data Sheet before handling.
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
Specifications
Media RecommendationExpress Five™ SFM (Serum-Free Media)
No. of Cells3 x 106 cells
Product LineHigh Five
Product TypeHigh Five Cells
Quantity3 x 106 cells
Cell LineHigh Five
Cell TypeInsect Cells
SpeciesTrichoplusia ni
Unit SizeEach
Contents & Storage
High Five Frozen Cells:
• Storage conditions: liquid nitrogen (vapor phase)

Express Five Serum-free Medium:
• Storage conditions: 2°–8°C
• Protect from light

L-glutamine, 100X:
• Storage conditions: -5 to -20°C
• Protect from light

Frequently asked questions (FAQs)

What is the procedure to thaw frozen insect cells?

The following protocol describes a general procedure for thawing cryopreserved cells. For detailed protocols, always refer to the cell-specific product insert.

1. Remove the cryovial containing the frozen cells from liquid nitrogen storage and immediately place it into a 37°C water bath.
2. Quickly thaw the cells (< 1 minute) by gently swirling the vial in the 37°C water bath until there is just a small bit of ice left in the vial.
3. Transfer the vial into a laminar flow hood. Before opening, wipe the outside of the vial with 70% ethanol.
4. Transfer the desired amount of pre-warmed complete growth medium appropriate for your cell line dropwise into the centrifuge tube containing the thawed cells.
5. Centrifuge the cell suspension at approximately 200 x g for 5-10 minutes. The actual centrifugation speed and duration varies depending on the cell type.
6. After the centrifugation, check the clarity of supernatant and visibility of a complete pellet. Aseptically decant the supernatant without disturbing the cell pellet.
7. Gently resuspend the cells in complete growth medium, and transfer them into the appropriate culture vessel and into the recommended culture environment.

Note: The appropriate flask size depends on the number of cells frozen in the cryovial, and the culture environment varies based on the cell and media type.

Why does the Insect cell line manual state: "Cells should be maintained at 27 degrees C in a non-humidified environment."

Insect cells do not require CO2 or high humidity to grow, they can grow in a lab drawer at room temperature. We recommend this so people don't waste CO2 and other resources necessary for maintaining a tissue culture incubator. It should be noted, however, that the cells will grow in a humidified incubator.

How does one get High Five cells that have been adapted for suspension culture to adhere to flasks? This can be important when generating stable cell lines with High Five cells.

To get High Five cells that have been adapted for suspension culture to adhere to a flask for selection of stable clones, it is recommended to check for heparin in the media (heparin helps keep the cells in suspension) and add 0.1% FBS to the culture for 10-20 minutes, then change the media. Keep in mind that cells which have adapted to suspension culture often take time to revert to adherent culture. Maintain the cells in the same flask over a few passages under selection, each time passaging at 1:4 or 1:5, without spinning the cells down. This should select for the adherent cells.

At what density should High Five cells be frozen?

We recommend a density of 3.0x10E6 cells/vial for adherent cultures and 10-15x10E6 cells/vial for suspension cultures.

Why do I need to add 18 mM L-Glutamine to prepare High Five Cells in Express Five Medium (Cat. No. B85502) and Express Five SFM (Cat. No. 10486025)?

L-Glutamine is an essential nutrient in cell cultures for energy production as well as protein and nucleic acid synthesis. In general, insect cell media contain higher L-Glutamine levels than serum-free media for mammalian cells, primary cells, stem cells, etc. This is to support the fast metabolism of insect cells, especially when used for virus production and protein expression applications.

Note that removal of L-Glutamine from the media formulation aids in product shelf-life by preventing glutamine degradation during storage.

Citations & References (35)

Citations & References
Abstract
OX40 stimulation by gp34/OX40 ligand enhances productive human immunodeficiency virus type 1 infection.
Authors: Takahashi Y; Tanaka Y; Yamashita A; Koyanagi Y; Nakamura M; Yamamoto N;
Journal:J Virol
PubMed ID:11435553
'OX40 is a member of the tumor necrosis factor (TNF) receptor superfamily and known to be an important costimulatory molecule expressed on activated T cells. To investigate the role of costimulation of OX40 in human immunodeficiency virus type 1 (HIV-1) infection by its natural ligand, gp34, the OX40-transfected ACH-2 cell ... More
Kinesin Superfamily Motor Protein KIF17 and mLin-10 in NMDA Receptor-Containing Vesicle Transport
Authors:Mitsutoshi Setou, Terunaga Nakagawa, Dae-Hyun Seog, Nobutaka Hirokawa *
Journal:Science
PubMed ID:10846156
'Experiments with vesicles containing N-methyl-D-aspartate (NMDA)receptor 2B (NR2B subunit) show that they are transported alongmicrotubules by KIF17, a neuron-specific molecular motor in neuronaldendrites. Selective transport is accomplished by direct interaction of theKIF17 tail with a PDZ domain of mLin-10 (Mint1/X11), which is aconstituent of a large protein complex including mLin-2 ... More
Alphaherpesvirus origin-binding protein homolog encoded by human herpesvirus 6B, a betaherpesvirus, binds to nucleotide sequences that are similar to ori regions of alphaherpesviruses.
Authors:Inoue N, Dambaugh TR, Rapp JC, Pellett PE
Journal:J Virol
PubMed ID:8207791
'We previously identified a human herpesvirus 6B (HHV-6B) homolog of the alphaherpesvirus origin-binding protein (OBP), exemplified by the herpes simplex virus type 1 UL9 gene product. This finding is of particular interest because HHV-6B is otherwise more closely related to members of the betaherpesvirus subfamily. The prototypic betaherpesvirus, human cytomegalovirus, ... More
Interaction of human nuclear topoisomerase I with guanosine quartet-forming and guanosine-rich single-stranded DNA and RNA oligonucleotides.
Authors: Marchand Christophe; Pourquier Philippe; Laco Gary S; Jing Naijie; Pommier Yves;
Journal:J Biol Chem
PubMed ID:11756434
'Human nuclear DNA topoisomerase I (top1) plays a crucial role in DNA replication, transcription, and chromosome condensation. In this study, we show that intra- and intermolecular guanosine quartets (G-quartets) can inhibit top1-mediated DNA cleavage at a high affinity site. Top1-mediated DNA cleavage was also inhibited by a 16-mer single-stranded oligodeoxynucleotide ... More
Purification and characterization of human DNA damage checkpoint Rad complexes.
Authors: Lindsey-Boltz L A; Bermudez V P; Hurwitz J; Sancar A;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11572977
'Checkpoint Rad proteins function early in the DNA damage checkpoint signaling cascade to arrest cell cycle progression in response to DNA damage. This checkpoint ensures the transmission of an intact genetic complement to daughter cells. To learn about the damage sensor function of the human checkpoint Rad proteins, we purified ... More