Thermo Scientific™
Shandon™ CytoRich™ Red Collection Fluid
 Related applications:
Anatomical Pathology
Allow for more accurate evaluation of cytological features with Thermo Scientific™ Shandon™ CytoRich™ Red Collection Fluid, which lyses RBCs and solubilizes proteins.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Pre-filled containers available
  • Ready-to-use, alcohol-based fluid
  • Compatible with immunohistochemistry
Documents
Manuals & protocols