Guanidine hydrochloride, NF, LBLE, GMP, Excipient Grade
Guanidine hydrochloride, NF, LBLE, GMP, Excipient Grade
BioSpectra

Guanidine hydrochloride, NF, LBLE, GMP, Excipient Grade

This item is suitable for use as an excipient, and is manufactured in accordance with the ICH-Q7 Good Manufacturing PracticeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberdimQuantity
BGHCL-3250-25
BGHCL-3250-10
BGHCL-3250-50
Catalog number BGHCL-3250-25
Price (KRW)
-
Request A Quote
This item is suitable for use as an excipient, and is manufactured in accordance with the ICH-Q7 Good Manufacturing Practice Guide. This grade of guanidine hydrochloride is not suitable to be used as an active pharmaceutical ingredient, drug product, or household item.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
FormCrystals
Molecular Weight (g/mol)95.53 g/mol
Packaging TypeHDPE round pail with lid, LDPE liner
Purity99.5 to 101%
Quantity25 kg
GradeNF
pH Range4.5 to 6.0 (6M)
Unit Size25 kg