VetMAX™ B. hyodysenteriae Kit
Applied Biosystems™

VetMAX™ B. hyodysenteriae Kit

Note: The name of this product has changed. It was previously called LSI VetMAX Brachyspira Hyodysenteriae. Same as TaqVet BrachyspiraLeia mais
Have Questions?
Número do catálogoQuantity
BHYO5050 tests
Número do catálogo BHYO50
Preço (BRL)
-
Quantity:
50 tests
Note: The name of this product has changed. It was previously called LSI VetMAX Brachyspira Hyodysenteriae.

Same as TaqVet Brachyspira hyodysenteriae Real-Time PCR Kit (BHYO/50)

TaqMan™ probes - DNA - duplex - One well reaction

Real-time PCR enables sensitive and specific detection of pathogen nucleic acid in animal samples, allowing for reliable and rapid screening and detection of infected animals.

The LSI VetMAX™ kits include:

• 1 ready-to-use mix that includes a set of oligonucleotides for detection of the target gene OR these separate components that will need to be mixed together: a set of oligonucleotides for the detection of the target gene + a TaqMan™ real-time PCR master mix + an RT enzyme (only for RNA kits).
• 1 external positive control of the same nature as the target (inactivated bacteria or virus). Use as a quality control for PCR and/or extraction yield.
• 1 internal positive control (exogenous) to be added at the time of extraction. Use as a quality control for extraction yield.
• A quality control certificate.

For veterinary use only.

Regulatory requirements vary by country; products may not be available in your geographic area.

Biological hazard safety

WARNING! BIOHAZARD. Biological samples such as tissues, body fluids, infectious agents, and blood of humans and other animals have the potential to transmit infectious diseases. All work should be conducted in properly equipped facilities using the appropriate safety equipment (for example, physical containment devices). Safety equipment also may include items for personal protection, such as gloves, coats, gowns, shoe covers, boots, respirators, face shields, safety glasses, or goggles. Individuals should be trained according to applicable regulatory and company/institution requirements before working with potentially biohazardous materials. Follow all applicable local, state/provincial, and/or national regulations. The following references provide general guidelines when handling biological samples in laboratory environment.

U.S. Department of Health and Human Services, Biosafety in Microbiological and Biomedical Laboratories (BMBL), 5th Edition, HHS Publication No. (CDC) 21-1112, Revised December 2009

World Health Organization, Laboratory Biosafety Manual, 3rd Edition, WHO/CDS/CSR/LYO/2004.11
Especificações
No. of Tests50 tests
Product LineVetMAX
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity50 tests
Sample TypeFeces
SpeciesSwine/Porcine, Porcine
Target Organism ClassB. hyodysenteriae
Detection MethodPrimer-probe
For Use With (Application)Real Time PCR (qPCR)
GC-Rich PCR PerformanceLow
Unit SizeEach