Search
View additional product information for Human IL-23 ELISA Kit - FAQs (BMS2023-3TEN, BMS2023-3)
18 product FAQs found
You can find expected "normal levels" listed in the Platinum ELISA Kit manual under "Expected Values". Please be aware that these normal values were obtained in our laboratories with a limited number of samples. We strongly suggest that each laboratory perform studies in order to establish normal range for their sample populations.
In case of small reagent volumes, please spin down vial before use to make sure to all contents are collected in the bottom of the tube.
We recommend following the instructions for sample dilutions as provided in the manual. We cannot guarantee optimal performance of the assay if internal assay dilutions as instructed were omitted.
Incubation times should be followed exactly as stated in the product manual to ensure optimal Platinum ELISA test performance.
An initial washing step is not necessary but recommended as accuracy may be increased.
We recommend that you use the manual included with the Platinum ELISA kit.
The reagents within the Platinum ELISA kits are not interchangeable between different lots of the same kit or between kits against different analytes.
No, but we strongly recommend that you make at least double determinations of each sample and standard point.
Most ELISAs are suitable for serum, plasma, cell culture supernatants and other body fluids. Detailed information is given in the product manual for each individual kit.
All reagents necessary to perform 1 or 10 plate ELISA assays, including optimized capture-antibody pre-coated plates.
The supernatants can be harvested and stored -20 degrees C. Avoid freeze/thaw cycles. We recommend that you briefly centrifuge samples after collection to pellet any dead/floating cells remaining in the supernatant. Transfer to new tube for storage.
Each Ready-Set-Go! ELISA Set comes with its own Certificate of Analysis (CofA). The directions on the CofA should be strictly followed to ensure performance of the reagents. The CofA gives specific instructions on how to dilute each component of your lot specific set. The instructions on the CofA should not be deviated from in order to ensure accurate results.
The calculation factor is always stated in the manual.
For some Instant ELISA kits, the reconstitution volume of the standard and blank wells may differ (these components are lot specific). Please use the manual provided in the kit for specific volumes to use.
In general, you can use undiluted samples with the Instant ELISA. Please refer to the product manual to determine if dilution of the sample is required for your kit of interest. The volume of sample required for the individual kits will also be listed in the product manual.
No, the secondary antibody is included as a component of the lyophilized reagents in the wells.
No, the recombinant standard is included as a component of the lyophilized reagents in the wells.
Performance of Platinum ELISA cannot be guaranteed if reagents are not stored as indicated.