IL-2 Human ELISA Kit, High Sensitivity - FAQs
IL-2 Human ELISA Kit, High Sensitivity - FAQs
View additional product information for IL-2 Human ELISA Kit, High Sensitivity - FAQs (BMS221HS)
5 product FAQs found
How should I prepare the standards for use with the High Sensitivity ELISA Kits?
We recommend that you prepare the standard in Eppendorf tubes.
Are the wash steps necessary when using your eBioscience High Sensitivity ELISA Kits?
Yes, please follow the washing steps exactly as outlined in the product manual (for optimal readings: an automatic plate washer is recommended).
For the eBioscience High Sensitivity ELISA Kits, can I perform the incubations at higher temperatures?
Incubations should occur at ambient temperatures not higher than 25 degrees C.
For the eBioscience High Sensitivity ELISA Kits, can I deviate from the recommended incubation times?
No, after adding the amplification reagents to the sample wells, the incubation times have to be followed exactly for optimal use.
For the eBioscience High Sensitivity ELISA Kits, can the amplification reagents be prepared ahead of time?
No, the amplification reagents have to be prepared immediately before use. The reagent AR1 contains methanol which evaporates easily.