Search
View additional product information for Human IL-1 beta ELISA Kit - FAQs (BMS224-2TEN, BMS224-2)
20 product FAQs found
Unfortunately, we do not offer the standards in this kit as a standalone product.
The IL-1 beta Human ELISA Kit comes with 2 vials of standards that do not need to be reconstituted at the same time. You can store the other vial of standard at -20 degrees C until needed.
The other vial of standard can be reconstituted and then aliquoted to allow for multiple use. The volume to reconstitute is written on the vial itself (and changes lot to lot), but you would want to aliquot into micro-tubes with just enough volume plus with a little extra (to avoid volume loss) to begin preparing your standard curve.
For example, you add 225 µl of the reconstituted standard into an Eppendorf tube which gets serially diluted and is enough to prepare duplicates for the standard curve.
Therefore all you need is 250 µl (225 µl+25 µl wiggle room/volume loss) of the reconstituted standard as an aliquot. Store the aliquots at minimum -20 degrees C.
This will allow you to run the assay a minimum of 2x (2 vials of standard) or more if you aliquot as discussed above.
Keep in mind the kit only comes with 12 strips. You will use 2 strips each time you want to run the assay for the standards alone (as they should be run in duplicates), as you cannot reuse the standard curve/OD from one run of the assay for another set. Similarly, you would need 2 strips per set of samples/unknowns to run in duplicate.
So, at most this kit would allow 3 uses. (4 strips per use, 2 for standards, 2 for unknowns).
Further, should you wish to use these in 3 separate occasions, be sure to allow the foil bag to come to room temperature (to avoid condensation issues), remove the strips from the holder that you don’t need and place these back into the bag and immediately return the foil bag back to 4 degrees C with the desiccant in place.
We do recommend storing your samples at -20 degrees C until you have collected enough to run the assay as indicated.
You can find expected "normal levels" listed in the Platinum ELISA Kit manual under "Expected Values". Please be aware that these normal values were obtained in our laboratories with a limited number of samples. We strongly suggest that each laboratory perform studies in order to establish normal range for their sample populations.
In case of small reagent volumes, please spin down vial before use to make sure to all contents are collected in the bottom of the tube.
We recommend following the instructions for sample dilutions as provided in the manual. We cannot guarantee optimal performance of the assay if internal assay dilutions as instructed were omitted.
Incubation times should be followed exactly as stated in the product manual to ensure optimal Platinum ELISA test performance.
An initial washing step is not necessary but recommended as accuracy may be increased.
We recommend that you use the manual included with the Platinum ELISA kit.
The reagents within the Platinum ELISA kits are not interchangeable between different lots of the same kit or between kits against different analytes.
No, but we strongly recommend that you make at least double determinations of each sample and standard point.
Most ELISAs are suitable for serum, plasma, cell culture supernatants and other body fluids. Detailed information is given in the product manual for each individual kit.
All reagents necessary to perform 1 or 10 plate ELISA assays, including optimized capture-antibody pre-coated plates.
The supernatants can be harvested and stored -20 degrees C. Avoid freeze/thaw cycles. We recommend that you briefly centrifuge samples after collection to pellet any dead/floating cells remaining in the supernatant. Transfer to new tube for storage.
Each Ready-Set-Go! ELISA Set comes with its own Certificate of Analysis (CofA). The directions on the CofA should be strictly followed to ensure performance of the reagents. The CofA gives specific instructions on how to dilute each component of your lot specific set. The instructions on the CofA should not be deviated from in order to ensure accurate results.
The calculation factor is always stated in the manual.
For some Instant ELISA kits, the reconstitution volume of the standard and blank wells may differ (these components are lot specific). Please use the manual provided in the kit for specific volumes to use.
In general, you can use undiluted samples with the Instant ELISA. Please refer to the product manual to determine if dilution of the sample is required for your kit of interest. The volume of sample required for the individual kits will also be listed in the product manual.
No, the secondary antibody is included as a component of the lyophilized reagents in the wells.
No, the recombinant standard is included as a component of the lyophilized reagents in the wells.
Performance of Platinum ELISA cannot be guaranteed if reagents are not stored as indicated.