Will IL-1 beta Human ELISA Kit, High Sensitivity (Cat. No. BMS224HS) detect both total and active IL1 beta?
The antibody pair in IL-1 beta Human ELISA Kit, High Sensitivity is designed to recognize the active form of Human IL-1 beta. The level of cross-reactivity for the inactive form has not been determined.