Product Image
Invitrogen™

eBioscience™ Annexin V Apoptosis Detection Kits

Kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량여기/방출콘주게이트
BMS500BT-100
BMS500BT/100으로도 사용됨
100 TestsNA, 535/NA, 617Biotin, Propidium Iodide
88-8005-74200 Tests488, 535/520, 617FITC, Propidium Iodide
88-8006-7250 Tests405, 546/450, 647eFluor 450, 7-AAD
88-8006-74200 Tests405, 546/450, 647eFluor 450, 7-AAD
88-8007-74200 Tests650, 535/660, 617APC, Propidium Iodide
88-8008-7250 TestsPCP-eFluor 710
88-8008-74200 TestPCP-eFluor 710
BMS500FI-100100 Tests488, 535/520, 617FITC, Propidium Iodide
BMS500FI-2020 Tests488, 535/520, 617FITC, Propidium Iodide
BMS500FI-300300 Tests488, 535/520, 617FITC, Propidium Iodide
88-8102-7250 TestsPE, 7-AAD
88-8102-74200 TestsPE, 7-AAD
88-8103-7250 TestsPE/Cy7
88-8103-74200 TestsPE/Cy7
BMS500BT-20
BMS500BT/20으로도 사용됨
20 TestsNA, 535/NA, 617Biotin, Propidium Iodide
BMS500BT-300
BMS500BT/300으로도 사용됨
300 TestsNA, 535/NA, 617Biotin, Propidium Iodide
88-8005-7250 Tests488, 535/520, 617FITC, Propidium Iodide
88-8007-7250 Tests650, 535/660, 617APC, Propidium Iodide
카탈로그 번호 BMS500BT-100
BMS500BT/100으로도 사용됨
제품 가격(KRW)
352,000
온라인 행사
Ends: 30-Sep-2026
414,000
할인액 62,000 (15%)
Each
수량:
100 Tests
여기/방출:
NA, 535/NA, 617
콘주게이트:
Biotin, Propidium Iodide
제품 가격(KRW)
352,000
온라인 행사
Ends: 30-Sep-2026
414,000
할인액 62,000 (15%)
Each
Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.
Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).

Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

For Research Use Only. Not for use in diagnostic procedures.
사양
설명eBioscience Annexin V-Biotin Apoptosis Detection Kit
여기/방출NA, 535/NA, 617
용도(장비)Flow Cytometer
제품 유형Apoptosis Detection Kit
수량100 Tests
배송 조건Wet Ice
콘주게이트Biotin, Propidium Iodide
Unit SizeEach
구성 및 보관
• Binding Buffer (4X), 1 bottle (50 mL)
• rh Annexin V-Biotin, 1 vial (0.5 mL)
• Propidium Iodide (20 μg/mL), 1 vial (1.8 mL). 2°C to 8°C. Avoid temperature fluctuations.

자주 묻는 질문(FAQ)

Can I use eFluor 450-conjugated antibodies with eBioscience Annexin V Apoptosis Detection kits?

Yes, eFluor 450-conjugated antibodies are compatible with eBioscience Annexin V Apoptosis Detection kits.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

인용 및 참조 문헌 (1)

인용 및 참조 문헌
Abstract
Pro-apoptotic and anti-proliferative effects of
Authors:Chairissy MD, Wulandari LR, Sujuti H
Journal:Int J Ophthalmol
PubMed ID:31544034
'To investigate the effect of'