eBioscience™ Annexin V-FITC Apoptosis Detection Kit, 300 Tests - FAQs
eBioscience™ Annexin V-FITC Apoptosis Detection Kit, 300 Tests - FAQs
View additional product information for eBioscience™ Annexin V Apoptosis Detection Kits - FAQs (88-8005-74, BMS500BT-100, 88-8103-72, BMS500BT-300, BMS500FI-20, 88-8102-72, BMS500BT-20, BMS500FI-100, 88-8103-74, 88-8102-74, BMS500FI-300, 88-8008-72, 88-8006-72, 88-8007-72, 88-8006-74, 88-8005-72, 88-8007-74, 88-8008-74)
1 product FAQs found
Can I use eFluor 450-conjugated antibodies with eBioscience Annexin V Apoptosis Detection kits?
Yes, eFluor 450-conjugated antibodies are compatible with eBioscience Annexin V Apoptosis Detection kits.