Monkey ICAM-1 (Soluble) Instant ELISA™ Kit - FAQs
Monkey ICAM-1 (Soluble) Instant ELISA™ Kit - FAQs
View additional product information for Monkey ICAM-1 (Soluble) Instant ELISA™ Kit - FAQs (BMS648INST)
1 product FAQs found
How many tests can I perform with an Instant ELISA Kit?
With an Instant ELISA, 128 tests (includes samples plus standards) can be performed. The number of tests results from the following composition:
One 96 well plate designed only for samples; 96 samples
Two 8-point standard curve in duplicates; 32 wells; Thus the kit can be used at two different time points, each with their own standard curve.