El kit de tampón de desplazamiento NativePAGE™ contiene el tampón de desplazamiento NativePAGE™ (20X), que se utiliza para elaborar losMás información
Have Questions?
Número de catálogo
Cantidad
BN2007
1 Kit
Número de catálogo BN2007
Precio (MXN)
-
Cantidad:
1 Kit
El kit de tampón de desplazamiento NativePAGE™ contiene el tampón de desplazamiento NativePAGE™ (20X), que se utiliza para elaborar los tampones de desplazamiento de cátodos y ánodos NativePAGE™ para su uso con una minicelda XCell SureLock® cuando se utilizan los geles NativePAGE™. El kit también contiene el aditivo de tampón de cátodos NativePAGE™ (20X) que se utiliza con el tampón de desplazamiento NativePAGE™ para crear el tampón de desplazamiento de cátodos para el sistema.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
TampónTampones de procesamiento, Tampones de desplazamiento
Compatibilidad del gelGeles NativePAGE™
Cantidad1 Kit
Tipo de gelGel Native
Línea de productosNativePAGE
Tipo de productoKit de tampones de desplazamiento PAGE
Unit SizeEach
Contenido y almacenamiento
Contiene: • 1 l de tampón de desarrollo NativePAGE™ (20X) • 250 ml de aditivo para tampón de cátodo NativePAGE™ (20X) Almacenar a temperatura ambiente.
Preguntas frecuentes
I would like to run a NativePAGE gel. Which of your protein standards should I use?
We recommend using the NativeMark Unstained Protein Standard, Cat. No. LC0725 for native gel electrophoresis with Tris-Glycine, NuPAGE Tris-Acetate or NativePAGE gels.
Can I use the NativePAGE Sample and Running buffers with NuPAGE Bis-Tris gels?
We do not recommend using NativePAGE Sample and Running buffers with NuPAGE Bis-Tris gels. NuPAGE Invitrogen Bis-Tris gels are optimized for denaturing conditions and have an extremely low operating pH (pH 7.0), which makes it difficult for most proteins to migrate through them in a native state.
Can I use the NativePAGE Sample and Running buffers with Invitrogen Tris-Glycine gels or NuPAGE Tris-Acetate gels for native applications?
NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate or Invitrogen Tris-Glycine gels. For those gels, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.
Can I use the NativePAGE Sample and Running buffers with Invitrogen Tris-Glycine gels for native applications?
NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including Invitrogen Tris-Glycine gels. For those gels, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.
Can I use the NativePAGE Sample and Running buffers with NuPAGE Tris-Acetate gels for native applications?
NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate gels. When running Tris-Acetate gels under native conditions, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.