Sodium hydroxide solution, 10N (30%), GMP
BioSpectra

Sodium hydroxide solution, 10N (30%), GMP

This item is suitable for use as a process chemical and is manufactured in accordance with the IPEC-PQG Joint Good Manufacturing Practice Guide.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
3.8 L
10 L
19 L
6 x 1 L
200 L
940 L
Número de catálogo BNH4150-CASE
Precio (MXN)
-
Cantidad:
6 x 1 L
This item is suitable for use as a process chemical and is manufactured in accordance with the IPEC-PQG Joint Good Manufacturing Practice Guide.
Low chloride, low iron, BET-tested, GMP manufactured bio pharma grade sodium hydroxide with WFI, this item is suitable for use as a process chemical. It is manufactured in accordance with the IPEC-PQG Joint Good Manufacturing Practice Guide. This grade of sodium hydroxide solution is not suitable to be used as an active pharmaceutical ingredient, drug product, or household item.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición105–142°C
CAS1310-73-2
Nombre del producto químico o materialSodium Hydroxide
ColorColorless
Fórmula molecularNaOH
EnvaseNon-fluorinated bottle with lid
SinónimoCaustic Soda, Lye, Sodium Hydrate
Peso molecular (g/mol)40
Cantidad6 x 1 L
pH14
Unit Size6 x 1 L