Cooked Meat Medium (Bottled)
Thermo Scientific™

Cooked Meat Medium (Bottled)

Grow and maintain aerobic and anaerobic organisms with these nutritious, ready prepared bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
BO0212E24/Pk.
EB0212E100/Pk.
Catalog number BO0212E
Price (MXN)
-
Quantity:
24/Pk.

Grow and maintain aerobic and anaerobic organisms with Thermo Scientific ™ Oxoid™ Cooked Meat Medium ready prepared bottles. The medium contains heart tissue, beef extracts, peptone and glucose for the cultivation of anaerobic and aerobic organisms. It has the ability to initiate bacterial growth from very small inocula and to maintain the viability of cultures over long periods of time1. Mixed cultures of bacteria survive in Cooked Meat Medium without displacing the slower growing organisms.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared bottles of Cooked Meat Medium.

  • Suitable for primary growth and maintenance of aerobic and anaerobic organisms.
  • Preserves the viability of organisms over a long period of time and is useful in maintaining anaerobic stock organisms.
  • Convenient: the pre-filled bottles are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months

The addition of glucose to the formulation allows rapid, heavy growth of anaerobic bacteria in a short time and leads to a more rapid identification of important anaerobes. The improved growth also enhances GLC identification of anaerobic bacteria.

The improved clarity of the supernatant broth permits earlier detection of growth especially when combined with the increased growth of most organisms. Slower growing isolates will yield detectable growth within 45 hours of incubation.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

1.Robertson M. (1916) J. Path. Bact. 20. 327-349.

Specifications
ColorStraw, slight haze
DescriptionCooked Meat Medium (Bottled)
Fill Weight10.0g + 0.3g
FormatUniversal Bottles
Packaging TypeTwenty four universal bottles with screw cap closures in a box
Quantity24/Pk.
Shelf Life52 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2 to 25°C