Nutrient Agar (Slope)
Thermo Scientific™

Nutrient Agar (Slope)

Cultivate non-fastidious microorganisms for testing food, milk and dairy products using these nutritious, ready prepared slopes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFill WeightPackaging Type
BO0336B24/Pk.3.0g + 0.3gTwenty four bijou bottles with screw cap closures in a box
EB0336B200/Pk.3.0g + 0.3gTwo hundred bijou bottles with screw cap closures in a box.
BO0336E24/Pk.10.0g + 0.3gTwenty four universal bottles with screw cap closures in a box.
Catalog number BO0336B
Price (MXN)
-
Quantity:
24/Pk.
Fill Weight:
3.0g + 0.3g
Packaging Type:
Twenty four bijou bottles with screw cap closures in a box

Cultivate non-fastidious microorganisms for testing food, milk and dairy products with Thermo Scientific™ Oxoid™ Nutrient Broth Slopes. This non-selective, fluid medium contains beef extracts combined with peptone, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared Slopes of Nutrient Broth.

  • Suitable for general cultivation of less fasidious microorganisms.
  • Convenient: the pre-filled slopes are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ColorStraw, clear
DescriptionNutrient Agar (Slope)
Fill Weight3.0g + 0.3g
FormatBijou bottles
Packaging TypeTwenty four bijou bottles with screw cap closures in a box
Quantity24/Pk.
Shelf Life52 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2 to 25°C