Columbia Agar with Chocolated Horse Blood (Slope)
Thermo Scientific™

Columbia Agar with Chocolated Horse Blood (Slope)

Isolate Haemophilus spp. from clinical specimens using these ready prepared slopes.

Have Questions?
Catalog number BO0341B
Price (MXN)
-

Isolate Haemophilus spp. from clinical specimens with Thermo Scientific™ Oxoid™ Columbia Agar with Chocolated Horse Blood Slopes. Chocolated horse blood is added to the general purpose Columbia Agar Base to supply the required nutrients for the growth of Haemophilus.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared bottles of Columbia Agar with Chocolated Horse Blood Slopes.

  • Supports the growth of Haemophilus with chocolated blood providing the nutritional requirements.
  • Convenient: the pre-filled slopes are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
ColorBrown, opaque
DescriptionColumbia Agar with Chocolated Horse Blood (Slope)
Fill Weight3.0g + 0.3g
FormatBijou bottles
Packaging TypeTwo hundred bijou bottles with screw cap closures in a box
Quantity200/Pk.
Shelf Life26 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 10°C