Oxoid™ Tryptone Soya Broth (Bottled)
Actual product may vary
Thermo Scientific™

Oxoid™ Tryptone Soya Broth (Bottled)

Grow a wide variety of microprganisms using this medium in convenient ready prepared bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFill Weight
BO0351M100.0 - 101.5g
BO0282M100 to 101.5 g
BO0369E10.0g + 0.3g
BO0282Z450 to 454 g
BO0369M100.0g + 1.5 g
BO0351E10.01 to 0.3 g
BO0351G20.0 - 20.3 g
BO0351U90 to 90.5 g
EB0351W1000 to 1005 g
BO0351V500.0 - 504.0 g
Catalog number BO0351M
Price (CLP)
-
Fill Weight:
100.0 - 101.5g
Grow a wide variety of microorganisms with Thermo Scientific™ Tryptone Soya Broth ready-prepared bottled medium. The general purpose medium, containing two peptones, is suitable for the cultivation of both aerobes and anaerobes. It is specified in International Pharmacopoeia1,2,3,4 and food testing methods for the growth of a wide variety of organisms.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Tryptone Soya Broth.

  • Conforms to formulations detailed in International Pharmacopoeia.
  • Convenient: pre-filled bottles are ready for immediate use.
  • Simple: easy to use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand.

General References:

  1. British Pharmacopoeia 2005
  2. European Pharmacopoeia 6th Edition 2008
  3. The Japanese Pharmacopoeia JP 15 2006
  4. The United States Pharmacopoeia USP 32 2009
Specifications
ATCC®ARTG
Certifications/ComplianceEP/USP
ColorStraw, clear
Country of OriginUK
Fill Weight100.0 - 101.5g
For Use With (Application)General Purpose Bottled Media
Media TypePrepared Media
Packaging TypePk.
Quantity10 x 100 mL
Shelf Life6 months
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 8°C