Milk Plate Count Agar
Thermo Scientific™

Milk Plate Count Agar

Isolate and enumerate microorganisms in milk and dairy products with this medium while conforming to ISO standards.
Have Questions?
Catalog NumberFill Weight
BO0652T250 to 254 g
Catalog number BO0652T
Price (MXN)
-
Fill Weight:
250 to 254 g
Isolate and enumerate microorganisms in milk and dairy products with this medium while conforming to ISO standards using Milk Plate Count Agar.
Specifications
ColorStraw
Fill Weight250 to 254 g
Format300 mL Sirop Bottle
Packaging Type10 Sirop Bottles with screw cap closures in a box
Quantity10/Pk.
Shelf Life52 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 25°C