Tryptone Soya Broth (X2 Strength)
Thermo Scientific™

Tryptone Soya Broth (X2 Strength)

Perform sterilty testing of your Pharmacopeial articles using this medium.
Have Questions?
Catalog NumberFill Weight
BO0830V500 to 504 g
Catalog number BO0830V
Price (EUR)
-
Fill Weight:
500 to 504 g
Triple strength media are used when large dilution volumes are needed. The higher concentration ensures medium strength is not overly diluted when large sample volumes are added.
Specifications
ColorStraw
Fill Weight500 to 504 g
Format500 mL DIN bottle
Packaging Type10 DIN bottles with wide septum, injectable closures in a correx box.
Quantity10/Pk.
Shelf Life52 Weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 25°C