Product Image
Thermo Scientific™
Oxoid™ Prepared MacConkey Broth Bottled Media
Oxoid™ Prepared MacConkey Broth Bottled Media, 100 mL, containing neutral red for the detection of coliform organisms in water and milk examination.
Have Questions?
카탈로그 번호 BO1124M
제품 가격(KRW)
-
Oxoid™ Prepared MacConkey Broth Bottled Media, 100 mL, containing neutral red for the detection of coliform organisms in water and milk examination.
사양
제품 유형Microbiology Media
Unit SizeEach