Group B Strep Selective Enrichment Broth
Thermo Scientific™

Group B Strep Selective Enrichment Broth

Selectively enrich and recover Group B Streptococcus (Streptococcus agalactiae) from clinical specimens, such as vaginal or rectal swabs.
Have Questions?
Número de catálogoPeso de llenado
BO1338EDe 10,0 a 10,3 g
Número de catálogo BO1338E
Precio (MXN)
-
Peso de llenado:
De 10,0 a 10,3 g
Group B Strep Selective Enrichment Broth is used to selectively enrich and recover Group B Streptococcus (Streptococcus agalactiae) from clinical specimens, such as vaginal or rectal swabs.
Especificaciones
ColorStraw
Peso de llenadoDe 10,0 a 10,3 g
FormatoPlastic universal bottle
Tipo de empaquetado24 Plastic Universal Bottles with screw cap closures in a box
Cantidad24/Pk.
Duración de almacenamiento26 Weeks
Origen de fabricaciónUnited Kingdom
Tipo de productoMicrobiology Media
Unit SizeEach
Contenido y almacenamiento
2°C to 25°C