Sensititre™ Bovine/Porcine BOPO6F Vet AST Plate
Sensititre™ Bovine/Porcine BOPO6F Vet AST Plate
Sensititre™ Bovine/Porcine BOPO6F Vet AST Plate
Thermo Scientific™
Sensititre™ Bovine/Porcine BOPO6F Vet AST Plate
Perform accurate AST with a bovine/porcine-specific formulary containing food animal antimicrobials using this convenient plate.
Catalog NumberQuantity
BOPO6F10/Pk.
Catalog number BOPO6F
Price (JPY)
-
Quantity:
10/Pk.
Perform accurate AST with a bovine/porcine-specific formulary, containing food animal antimicrobials using the Thermo Scientific™ Sensititre™ Bovine/Porcine BOPO6F AST Plate. Sensititre Bovine/Porcine BOPO6F AST Plate delivers accurate results on a single plate format, reducing the need for offline tests and retests.
Achieve reliable, consistent results for non-fastidious Gram-positive and Gram-negative isolates of veterinary origin with Sensititre Bovine/Porcine BOPO6F Plate.
  • Extended shelf life, room temperature storage eliminates inventory control concerns.
  • Individual packaging allows the laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Automated or manual read options are available for ultimate flexibility
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
For Veterinary Use Only.
For Veterinary Use Only.
Specifications
Antibiotic CodeVarious
Antimicrobial AgentsVarious
DescriptionSensititre Bovine/Porcine MIC Plate
Isolates Per Plate1
Quantity10/Pk.
Reading OptionsAutoread or manual
Specimen TypeBovine/Porcine Isolates
Product TypeAST Plate
SpeciesBovine, Porcine
Target Organism ClassNon-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test TypeVeterinary AST, Minimum Inhibitory Concentration (MIC)
Unit SizeEach