Thermo Scientific™
BP19A/IBP19BD Beta Probes
Catalog number: BP19DD
Thermo Scientific™
BP19A/IBP19BD Beta Probes
Catalog number: BP19DD
Thermo Scientific™ BP19A/IBP19BD Beta Probes monitor beta radiation across the beta energy spectrum with good efficiency and high sensitivity. It is an easy to operate with a slim profile and low weight.
Catalog Number
BP19DD
Unit Size
Each
Description
Low energy Beta Probe
Connector Types
MHV connector
Price (USD)
Full specifications
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionLow energy Beta Probe
Window Area100cm²
Item DescriptionLow energy Beta; MHV connector
Connector TypesMHV connector
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Beta Probe
Connector Types
PET connector
Price (USD)
Full specifications
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionBeta Probe
Window Area100cm²
Item DescriptionPET connector
Connector TypesPET connector
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Beta Probe
Connector Types
MHV connector
Price (USD)
Full specifications
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionBeta Probe
Window Area100cm²
Item DescriptionMHV connector
Connector TypesMHV connector
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Ruggedized Probe
Connector Types
PET connector
Price (USD)
Full specifications
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionRuggedized Probe
Window Area100cm²
Item DescriptionRuggedized; PET connector
Connector TypesPET connector
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Ruggedized Probe; Low energy Beta
Connector Types
MHV connector
Price (USD)
Full specifications
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionRuggedized Probe; Low energy Beta
Window Area100cm²
Item DescriptionLow energy Beta; MHV; Ruggedized
Connector TypesMHV connector
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Beta Probe
Connector Types
Selectra only
Price (USD)
Full specifications
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionBeta Probe
Window Area100cm²
Item DescriptionSelectra only
Connector TypesSelectra only
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Low energy Beta Probe
Connector Types
Selectra only
Price (USD)
Full specifications
Radiation TypeBeta
TypeSmart Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionLow energy Beta Probe
Window Area100cm²
Item DescriptionLow energy Beta, Selectra only
Connector TypesSelectra only
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Ruggedized Probe
Connector Types
MHV connector
Price (USD)
Full specifications
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionRuggedized Probe
Window Area100cm²
Item DescriptionRuggedized; MHV connector
Connector TypesMHV connector
Unit SizeEach
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeDescriptionConnector TypesPrice (USD)
BP19DDFull specifications
EachLow energy Beta ProbeMHV connectorRequest A Quote
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionLow energy Beta Probe
Window Area100cm²
Item DescriptionLow energy Beta; MHV connector
Connector TypesMHV connector
Unit SizeEach
BP19ADFull specifications
EachBeta ProbePET connectorRequest A Quote
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionBeta Probe
Window Area100cm²
Item DescriptionPET connector
Connector TypesPET connector
Unit SizeEach
BP19BDFull specifications
EachBeta ProbeMHV connectorRequest A Quote
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionBeta Probe
Window Area100cm²
Item DescriptionMHV connector
Connector TypesMHV connector
Unit SizeEach
BP19RAFull specifications
EachRuggedized ProbePET connectorRequest A Quote
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionRuggedized Probe
Window Area100cm²
Item DescriptionRuggedized; PET connector
Connector TypesPET connector
Unit SizeEach
BP19RDFull specifications
EachRuggedized Probe; Low energy BetaMHV connectorRequest A Quote
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionRuggedized Probe; Low energy Beta
Window Area100cm²
Item DescriptionLow energy Beta; MHV; Ruggedized
Connector TypesMHV connector
Unit SizeEach
IBP19BDFull specifications
EachBeta ProbeSelectra onlyRequest A Quote
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionBeta Probe
Window Area100cm²
Item DescriptionSelectra only
Connector TypesSelectra only
Unit SizeEach
IBP19DDFull specifications
EachLow energy Beta ProbeSelectra onlyRequest A Quote
Radiation TypeBeta
TypeSmart Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionLow energy Beta Probe
Window Area100cm²
Item DescriptionLow energy Beta, Selectra only
Connector TypesSelectra only
Unit SizeEach
BP19RBFull specifications
EachRuggedized ProbeMHV connectorRequest A Quote
Radiation TypeBeta
TypeBeta Probe
Efficiency14% 14C, 32% 60Co, 49% 36Cl, 51% 90Sr/90Y (BP19AD); 21% 14C, 34% 60Co, 48% 36Cl, 51% 90Sr/90Y (BP19DD)
Weight (English)1.1 lb.
Weight (Metric)0.5 kg
Background Measurements-1: <10, <8 (BP19AD, BP19DD); cpm: <600, <480 (BP19AD, BP19DD)
DescriptionRuggedized Probe
Window Area100cm²
Item DescriptionRuggedized; MHV connector
Connector TypesMHV connector
Unit SizeEach
Showing 8 of 8
Particular emphasis is placed on lower background response and higher low energy beta response.
  • 100cm2 rectangular-shaped window
  • Large scintillation probe with a BC400 plastic phosphor in a light alloy diecast housing
Recommended for:
  • Used for general purpose and large area monitoring for low, medium, and high energy beta
  • IBP19BD is an Intelligent version for use with Selectra range rate meters
  • Endura phosphor version available
Figures