Oxoid™ Basic Fuchsin Indicator
Thermo Scientific™
Oxoid™ Basic Fuchsin Indicator
Oxoid Basic Fuchsin Indicator is a dye for use with Endo Agar Base, Part No. CM0479B and m Endo (LES) Agar Base, Part No. MM0551B.
Have Questions?
카탈로그 번호수량
BR0050A10 g
카탈로그 번호 BR0050A
제품 가격(KRW)
-
수량:
10 g
Oxoid Basic Fuchsin Indicator is a dye for use with Endo Agar Base, Part No. CM0479B and m Endo (LES) Agar Base, Part No. MM0551B.
사양
설명Basic Fuchsin Indicator
수량10 g
형태Powder
제품 유형Indicator Dye
Unit SizeEach