Consumables for ESI Switching Valves
Thermo Scientific™
Consumables for ESI Switching Valves
Maintain sample introduction consistency with reliable rotors and stators for ESI switching valves, built for smooth operation and consistent analytic performance.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipoMaterial
BRE0003835StatorCTFE
BRE0003834RotorPFA
Número de catálogo BRE0003835
Precio (USD)
-
Tipo:
Stator
Material:
CTFE
Accelerate workflows and reduce downtime with these consumables for the switching valves on ESI FAST and prepFAST systems, including precision rotors and stators that maintain leak‑tight sealing and smooth switching. Designed for automated sample introduction systems, these direct‑fit parts support consistent throughput and reproducible results during routine replacement.
Increase throughput
  • Enable rapid, reliable valve switching to keep autosamplers moving samples efficiently.
Maintain seal integrity
  • Preserve leak‑tight interfaces
  • Reduce carryover and rework
  • Install easily
Support stable sample delivery
  • Improve run‑to‑run reproducibility
  • Minimize downtime
Especificaciones
MaterialCTFE
Para utilizar con (equipo)ESI prepFAST 2, P7+ valve
TipoStator
Unit SizeEach