Torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Thermo Scientific™
Torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Get improved plasma stability from the tulip design and easy, push-in insertion of Thermo Scientific™ torches for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
BRE0013019ICP-MS Organic Torch
1230790ICP-MS Quartz Torch
BRE0024760ICP-MS PLUS Torch
Catalog number BRE0013019
Price (USD)
449.00
Each
Type:
ICP-MS Organic Torch
Price (USD)
449.00
Each
Gain improved plasma stability from the tulip design and easy, push-in Gain improved plasma stability from the tulip design and easy, push-in insertion of fully demountable, single piece, Thermo Scientific™ Torches with connection-free gas supply for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems. All connectivity (argon gas supplies and plasma ignition) is contained within the torch holder to reduce complexity and improve usability.
Specifications
For Use With (Equipment)iCAP MX and iCAP Q/Qnova Series ICP-MS Systems
TypeICP-MS Organic Torch
Unit SizeEach