Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Actual product may vary
Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System
Thermo Scientific™

Peristaltic Pump Tubing for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS System

Designed for durability and fitness for purpose, a range of Thermo Scientific™ Peristaltic Pump Tubing for the iCAP™ MX and iCAP™ Q/Qnova Series is available to suit your sample matrices and ICP-MS applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDiameter (Metric)Material
BRE0013427Peristaltic Pump Tubing0.508 mmSolvent Flex
1320040Peristaltic Pump Tubing0.381 mmPVC
1320050Peristaltic Pump Tubing0.508 mmPVC
1320060Peristaltic Pump Tubing0.762 mmPVC
BRE0013426Peristaltic Pump Tubing0.381 mmSolvent Flex
BRE0013429Peristaltic Pump Tubing1.295 mmSolvent Flex
BRE0013428Peristaltic Pump Tubing0.762 mmSolvent Flex
1320110Peristaltic Pump Tubing1.295 mmSantoprene
1320120Peristaltic Pump Tubing3.175 mmSantoprene
1329400Peristaltic Pump Tubing0.254 mmPVC, Orange/Black
Catalog number BRE0013427
Price (USD)
-
Type:
Peristaltic Pump Tubing
Diameter (Metric):
0.508 mm
Material:
Solvent Flex
Suit your sample matrices and applications with this comprehensive range of Thermo Scientific™ Peristaltic Pump Tubing for the Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS systems; all designed for durability and fitness for purpose.

Select High Quality Consumables for Improved Performance
Choose from the following Peristaltic Pump Tubing:

  • Peristaltic Pump Tubing - dia.0,508 -Orange/Yellow -PVC
  • Peristaltic Pump Tubing - dia.0,381 -Orange/Green - PVC
  • Peristaltic Pump Tubing - dia.0,762 - Black/Black -PVC
  • Peristaltic Pump Tubing - dia.0,254 - Orange/Blue -PVC
  • Peristaltic Pump Tubing - dia.1,295 - Gray/Gray -Santoprene
  • Peristaltic Pump Tubing - dia.3,175 - Black/White -Santoprene
Specifications
Diameter (Metric)0.508 mm
For Use With (Equipment)iCAP MX and iCAP Q/Qnova Series
MaterialSolvent Flex
TypePeristaltic Pump Tubing
Unit SizePack of 12