Teledyne CETAC ENC-DC Ventilated Autosampler Enclosures
Teledyne CETAC ENC-DC Ventilated Autosampler Enclosures
Teledyne CETAC ENC-DC Ventilated Autosampler Enclosures
Teledyne CETAC ENC-DC Ventilated Autosampler Enclosures
Thermo Scientific™
Teledyne CETAC ENC-DC Ventilated Autosampler Enclosures
Protect your samples and personnel from contamination and harmful vapors with Teledyne CETAC ENC-DC Series ventilated dust cover enclosures for autosamplers.
製品番号(カタログ番号)使用対象 (装置)タイプ内容
BRE0020245CETAC EXR-8/XLR-8 /XLR-860Autosampler EnclosureDust Cover
BRE0020243CETAC ASX-520/520HS/560Autosampler EnclosureDust Cover
BRE0020244CETAC ASX-260/280Autosampler EnclosureDust Cover
BRE0020246CETAC Ventilated Dust CoversHEPA FilterHEPA Filter, Adapter
BRE0020247CETAC ASX-560Autosampler Enclosure BaseEnclosure Base
BRE0020248CETAC ASX-280Autosampler Enclosure BaseEnclosure Base
BRE0020249CETAC XLR-860Autosampler Enclosure BaseEnclosure Base
BRE0020250CETAC Ventilated Dust CoversAutosampler Enclosure FanAustralian Plug
BRE0020251CETAC Ventilated Dust CoversAutosampler Enclosure Fan2-pin Plug (Europe)
BRE0020252CETAC Ventilated Dust CoversAutosampler Enclosure Fan2-pin Plug (North America)
製品番号(カタログ番号) BRE0020245
価格(JPY)
-
使用対象 (装置):
CETAC EXR-8/XLR-8 /XLR-860
タイプ:
Autosampler Enclosure
内容:
Dust Cover
Protect your samples and personnel from contamination and harmful vapors with Teledyne CETAC ENC-DC Series Ventilated Autosampler Enclosures. These enclosures are made from shatter-proof polycarbonate materials with multiple ports for fully integrated ventilation using either an active fan or your in-house exhaust extraction system. The enclosures feature an optional HEPA filter to maintain sample integrity.
Multiple configuration possibilities and optional accessories allow you to configure the enclosure to best suit your requirements.
Compatible with Teledyne CETAC ASX-280, ASX-560 and XLR-860 Autosamplers.
  • Multiple ports - Placement of the vent and fan and routing tubing and power cords can be configured for any space.
  • Filtered positive pressure enclosure - When fitted with the optional fan and HEPA filter, the enclosure becomes a positive pressure enclosure.
  • Vented enclosure - The addition of the flange allows the fumes to be removed through a laboratory exhaust system.
  • Dust cover - ENC-DC Enclosures can be used without the fan and filter to function as a convenient dust cover.
  • Rigid base - ENC-DC Enclosures can be fitted with an optional base for use on irregular surfaces.
仕様
概要Ventilated Dust Cover/Enclosure for Teledyne CETAC XLR-860 Autosampler
内容Dust Cover
使用対象 (装置)CETAC EXR-8/XLR-8 /XLR-860
タイプAutosampler Enclosure
Unit SizeEach