Autosampler Polypropylene Tubes
Autosampler Polypropylene Tubes
Thermo Scientific™
Autosampler Polypropylene Tubes
Available in a variety of sizes to match your selected autosampler rack.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
BRE003285350 mL Polypropylene Tubes, Pack of 10
9423470041418 mL Polypropylene Tubes, 13 mm x 100 mm
94234700416120 mL Polypropylene Tubes, 21.5 mm x 100 mm
94234700411130 mL Polypropylene Tubes, 25 mm x 95 mm
94234700413114 mL Polypropylene Tubes, 16 mm x 100 mm
94234700415150 mL Polypropylene Standard Tubes, 30 mm x 115 mm
BRE003285212 mL Polypropylene Tubes
Catalog number BRE0032853
Price (USD)/ Pack of 10
-
Request A Quote
Description:
50 mL Polypropylene Tubes, Pack of 10
Choose the right tubes for your autosampler. Autosampler polypropylene tubes are available in a variety of different sizes to match your selected autosampler rack.
Select Tube Sizes to Fit Your Laboratory's Sample Racks
  • 8 mL polypropylene tubes for 90-position rack (1000)
  • 14 mL polypropylene tubes for 60-position rack (1000)
  • 20 mL polypropylene tubes for 40-position rack (500)
  • 30 mL polypropylene tubes for 24-position rack (500)
  • 50 mL polypropylene tubes for standard rack and 21-position rack (500)
Specifications
TypeAutosampler Polypropylene Tubes
Description50 mL Polypropylene Tubes, Pack of 10
For Use With (Equipment)Standard Rack and 21-position Autosampler Racks
Unit SizePack of 10