iCAP™ PRO X ICP-OES Duo - FAQs
iCAP™ PRO X ICP-OES Duo - FAQs
View additional product information for iCAP™ PRO ICP-OES and iCAP™ PRO X ICP-OES - FAQs (BRE731403, BRE731402, BRE731401, BRE731400)
This product does not have any FAQs available.